Assay Details
| Gene Symbol: CG43707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273074.1 | NP_001260003.1 | 1 - 2 | 702 | 71 | |
| NM_001273075.1 | NP_001260004.1 | 1 - 2 | 702 | 71 | ||
| NM_001273076.1 | NP_001260005.1 | 1 - 2 | 702 | 71 | ||
| NM_001273077.2 | NP_001260006.1 | 1 - 2 | 702 | 71 | ||
| NM_001273078.1 | NP_001260007.1 | 1 - 2 | 702 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001273075.1 transcripts.
Set Membership: |
|

CG43707
Genomic Map