Assay Details
| Gene Symbol: Bsg25D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201769.2 | NP_001188698.1 | 3 - 4 | 560 | 128 | |
| NM_001201770.2 | NP_001188699.1 | 3 - 4 | 1080 | 128 | ||
| NM_001273163.2 | NP_001260092.1 | 3 - 4 | 1056 | 128 | ||
| NM_057568.6 | NP_476916.4 | 3 - 4 | 1056 | 128 | ||
| NM_164637.3 | NP_723072.1 | 3 - 4 | 1080 | 128 | ||
| NM_164638.3 | NP_723073.1 | 3 - 4 | 560 | 128 | ||
| GenBank mRNA | AY095187.1 | - | 3 - 4 | 1062 | 128 | |
| BT100004.1 | - | 3 - 4 | 557 | 128 | ||
More Information
Set Membership: |
|

Bsg25D
Genomic Map