Assay Details
| Gene Symbol: Bsg25D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201769.2 | NP_001188698.1 | 1 - 2 | 96 | 79 | |
| NM_164638.3 | NP_723073.1 | 1 - 2 | 96 | 79 | ||
| GenBank mRNA | BT100004.1 | - | 1 - 2 | 93 | 79 | |
More Information
Set Membership: |
|

Bsg25D
Genomic Map