Assay Details
| Gene Symbol: NimB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176031.3 | NP_788045.1 | 3 - 4 | 993 | 60 | |
| GenBank mRNA | AF160879.1 | - | 3 - 4 | 954 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NimB1| Gene Symbol: NimB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176031.3 | NP_788045.1 | 3 - 4 | 993 | 60 | |
| GenBank mRNA | AF160879.1 | - | 3 - 4 | 954 | 60 | |
Set Membership: |
|