Assay Details
| Gene Symbol: Debcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176098.3 | NP_788278.1 | 1 - 2 | 920 | 77 | |
| GenBank mRNA | AB032430.2 | - | 1 - 2 | 295 | 77 | |
| AF178430.1 | - | 1 - 2 | 705 | 77 | ||
| AF222004.1 | - | 1 - 2 | 832 | 77 | ||
| AF244352.1 | - | 1 - 2 | 840 | 77 | ||
| BT022157.1 | - | 1 - 2 | 920 | 77 | ||
More Information
Set Membership: |
|

Debcl
Genomic Map