Assay Details
| Gene Symbol: Debcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176098.3 | NP_788278.1 | 2 - 3 | 1068 | 65 | |
| GenBank mRNA | AB032430.2 | - | 2 - 3 | 443 | 65 | |
| AF178430.1 | - | 2 - 3 | 853 | 65 | ||
| AF222004.1 | - | 2 - 3 | 980 | 65 | ||
| AF244352.1 | - | 2 - 3 | 988 | 65 | ||
| BT022157.1 | - | 2 - 3 | 1068 | 65 | ||
More Information
Set Membership: |
|

Debcl
Genomic Map