Assay Details
| Gene Symbol: Pkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103767.2 | NP_001097237.1 | 3 - 4 | 1454 | 61 | |
| NM_001169624.1 | NP_001163095.1 | 3 - 4 | 1454 | 61 | ||
| NM_001201966.2 | NP_001188895.1 | 5 - 6 | 1839 | 61 | ||
| NM_001201967.1 | NP_001188896.1 | 3 - 4 | 1454 | 61 | ||
| NM_001201968.3 | NP_001188897.3 | 4 - 5 | 1359 | 61 | ||
| NM_001259291.2 | NP_001246220.1 | 4 - 5 | 1300 | 61 | ||
| NM_001259292.2 | NP_001246221.1 | 4 - 5 | 1431 | 61 | ||
| NM_001299312.1 | NP_001286241.1 | 4 - 5 | 1300 | 61 | ||
| NM_001299313.1 | NP_001286242.1 | 4 - 5 | 1300 | 61 | ||
| NM_001299314.1 | NP_001286243.1 | 4 - 5 | 1300 | 61 | ||
| NM_001299315.1 | NP_001286244.1 | 4 - 5 | 1300 | 61 | ||
| NM_176110.2 | NP_788290.1 | 4 - 5 | 1300 | 61 | ||
| NM_176111.2 | NP_788291.1 | 4 - 5 | 1300 | 61 | ||
| NM_176112.2 | NP_788292.1 | 4 - 5 | 1422 | 61 | ||
| NM_176113.3 | NP_788293.2 | 4 - 5 | 1422 | 61 | ||
| GenBank mRNA | BT133091.1 | - | 4 - 5 | 1263 | 61 | |
More Information
Set Membership: |
|

Pkn
Genomic Map