Assay Details
| Gene Symbol: Pkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103767.2 | NP_001097237.1 | 7 - 8 | 4400 | 64 | |
| NM_001169624.1 | NP_001163095.1 | 6 - 7 | 3749 | 64 | ||
| NM_001201966.2 | NP_001188895.1 | 9 - 10 | 4344 | 64 | ||
| NM_001201967.1 | NP_001188896.1 | 7 - 8 | 3959 | 64 | ||
| NM_001201968.3 | NP_001188897.3 | 8 - 9 | 3864 | 64 | ||
| NM_001259291.2 | NP_001246220.1 | 8 - 9 | 3805 | 64 | ||
| NM_001259292.2 | NP_001246221.1 | 8 - 9 | 3936 | 64 | ||
| NM_001299312.1 | NP_001286241.1 | 9 - 10 | 4456 | 64 | ||
| NM_001299313.1 | NP_001286242.1 | 8 - 9 | 3526 | 64 | ||
| NM_001299314.1 | NP_001286243.1 | 8 - 9 | 3865 | 64 | ||
| NM_001299315.1 | NP_001286244.1 | 8 - 9 | 3859 | 64 | ||
| NM_176110.2 | NP_788290.1 | 7 - 8 | 3595 | 64 | ||
| NM_176111.2 | NP_788291.1 | 8 - 9 | 4246 | 64 | ||
| NM_176112.2 | NP_788292.1 | 7 - 8 | 3717 | 64 | ||
| NM_176113.3 | NP_788293.2 | 8 - 9 | 4368 | 64 | ||
| NM_176114.3 | NP_788294.1 | 4 - 5 | 1465 | 64 | ||
| GenBank mRNA | BT133091.1 | - | 8 - 9 | 4209 | 64 | |
| BT133361.1 | - | 4 - 5 | 1474 | 64 | ||
More Information
Set Membership: |
|

Pkn
Genomic Map