Assay Details
| Gene Symbol: Pkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103767.2 | NP_001097237.1 | 9 - 10 | 4659 | 89 | |
| NM_001169624.1 | NP_001163095.1 | 8 - 9 | 4008 | 89 | ||
| NM_001201966.2 | NP_001188895.1 | 11 - 12 | 4603 | 89 | ||
| NM_001201967.1 | NP_001188896.1 | 9 - 10 | 4218 | 89 | ||
| NM_001201968.3 | NP_001188897.3 | 10 - 11 | 4123 | 89 | ||
| NM_001259291.2 | NP_001246220.1 | 10 - 11 | 4064 | 89 | ||
| NM_001259292.2 | NP_001246221.1 | 10 - 11 | 4195 | 89 | ||
| NM_001299312.1 | NP_001286241.1 | 11 - 12 | 4715 | 89 | ||
| NM_001299313.1 | NP_001286242.1 | 10 - 11 | 3785 | 89 | ||
| NM_001299314.1 | NP_001286243.1 | 10 - 11 | 4124 | 89 | ||
| NM_001299315.1 | NP_001286244.1 | 10 - 11 | 4118 | 89 | ||
| NM_176110.2 | NP_788290.1 | 9 - 10 | 3854 | 89 | ||
| NM_176111.2 | NP_788291.1 | 10 - 11 | 4505 | 89 | ||
| NM_176112.2 | NP_788292.1 | 9 - 10 | 3976 | 89 | ||
| NM_176113.3 | NP_788293.2 | 10 - 11 | 4627 | 89 | ||
| NM_176114.3 | NP_788294.1 | 6 - 7 | 1724 | 89 | ||
| GenBank mRNA | BT133091.1 | - | 10 - 11 | 4468 | 89 | |
| BT133361.1 | - | 6 - 7 | 1733 | 89 | ||
More Information
Set Membership: |
|

Pkn
Genomic Map