Assay Details
| Gene Symbol: Pkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103767.2 | NP_001097237.1 | 4 - 5 | 2762 | 69 | |
| NM_001299312.1 | NP_001286241.1 | 5 - 6 | 2608 | 69 | ||
| NM_176111.2 | NP_788291.1 | 5 - 6 | 2608 | 69 | ||
| NM_176113.3 | NP_788293.2 | 5 - 6 | 2730 | 69 | ||
| GenBank mRNA | BT133091.1 | - | 5 - 6 | 2571 | 69 | |
More Information
Set Membership: |
|

Pkn
Genomic Map