Assay Details
| Gene Symbol: Pkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103767.2 | NP_001097237.1 | 5 - 6 | 3411 | 56 | |
| NM_001299314.1 | NP_001286243.1 | 6 - 7 | 2876 | 56 | ||
| NM_001299315.1 | NP_001286244.1 | 6 - 7 | 2870 | 56 | ||
| NM_176111.2 | NP_788291.1 | 6 - 7 | 3257 | 56 | ||
| NM_176113.3 | NP_788293.2 | 6 - 7 | 3379 | 56 | ||
| GenBank mRNA | BT133091.1 | - | 6 - 7 | 3220 | 56 | |
More Information
Set Membership: |
|

Pkn
Genomic Map