Assay Details
| Gene Symbol: Pkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176114.3 | NP_788294.1 | 1 - 2 | 267 | 73 | |
| GenBank mRNA | BT133361.1 | - | 1 - 2 | 276 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pkn| Gene Symbol: Pkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176114.3 | NP_788294.1 | 1 - 2 | 267 | 73 | |
| GenBank mRNA | BT133361.1 | - | 1 - 2 | 276 | 73 | |
Set Membership: |
|