Assay Details
| Gene Symbol: KCNQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144159.2 | NP_001137631.1 | 3 - 4 | 559 | 77 | |
| NM_001144160.2 | NP_001137632.1 | 3 - 4 | 820 | 77 | ||
| NM_001273919.1 | NP_001260848.1 | 3 - 4 | 550 | 77 | ||
| NM_001273920.1 | NP_001260849.1 | 3 - 4 | 559 | 77 | ||
| NM_001273921.1 | NP_001260850.1 | 3 - 4 | 559 | 77 | ||
| NM_001273922.1 | NP_001260851.1 | 3 - 4 | 559 | 77 | ||
| NM_001273923.1 | NP_001260852.1 | 3 - 4 | 559 | 77 | ||
| NM_176119.5 | NP_788299.3 | 3 - 4 | 559 | 77 | ||
| NM_176120.5 | NP_788300.3 | 3 - 4 | 559 | 77 | ||
| GenBank mRNA | AY823300.1 | - | 3 - 4 | 553 | 77 | |
| AY823301.1 | - | 3 - 4 | 553 | 77 | ||
More Information
Set Membership: |
|

KCNQ
Genomic Map