Assay Details
| Gene Symbol: GalT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176148.2 | NP_788328.1 | 1 - 2 | 523 | 71 | |
| NM_176149.2 | NP_788329.1 | 1 - 2 | 523 | 71 | ||
| GenBank mRNA | AY071036.1 | - | 1 - 2 | 519 | 71 | |
More Information
Set Membership: |
|

GalT1
Genomic Map