Assay Details
| Gene Symbol: CG33012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176155.2 | NP_788335.2 | 4 - 5 | 1068 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG33012| Gene Symbol: CG33012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176155.2 | NP_788335.2 | 4 - 5 | 1068 | 63 | |
Set Membership: |
|