Assay Details
| Gene Symbol: s-cup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136948.3 | NP_610792.2 | 1 - 2 | 527 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

s-cup| Gene Symbol: s-cup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136948.3 | NP_610792.2 | 1 - 2 | 527 | 67 | |
Set Membership: |
|