Assay Details
| Gene Symbol: Ack-like | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274005.1 | NP_001260934.1 | 7 | 4764 | 120 | |
| NM_001274006.1 | NP_001260935.1 | 7 | 4764 | 120 | ||
More Information
Set Membership: |
|

Ack-like
Genomic Map