Assay Details
| Gene Symbol: Impbeta11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176177.2 | NP_788357.1 | 1 - 2 | 2198 | 85 | |
| GenBank mRNA | AY069650.1 | - | 1 - 2 | 2181 | 85 | |
More Information
Set Membership: |
|

Impbeta11
Genomic Map