Assay Details
| Gene Symbol: Mctp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043094.2 | NP_001036559.2 | 7 - 8 | 2117 | 93 | |
| NM_001043095.3 | NP_001036560.1 | 2 - 3 | 926 | 93 | ||
| NM_001274149.1 | NP_001261078.1 | 7 - 8 | 2225 | 93 | ||
| NM_001299661.1 | NP_001286590.1 | 7 - 8 | 2168 | 93 | ||
| NM_001316428.1 | NP_001303357.1 | 7 - 8 | 2225 | 93 | ||
| NM_137528.5 | NP_611372.3 | 7 - 8 | 2168 | 93 | ||
| GenBank mRNA | AY071153.1 | - | 2 - 3 | 915 | 93 | |
More Information
Set Membership: |
|

Mctp
Genomic Map