Assay Details
| Gene Symbol: Mctp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043094.2 | NP_001036559.2 | 10 - 11 | 2799 | 76 | |
| NM_001043095.3 | NP_001036560.1 | 5 - 6 | 1608 | 76 | ||
| NM_001274149.1 | NP_001261078.1 | 10 - 11 | 2907 | 76 | ||
| NM_001299661.1 | NP_001286590.1 | 10 - 11 | 2850 | 76 | ||
| NM_001316428.1 | NP_001303357.1 | 10 - 11 | 2907 | 76 | ||
| NM_137528.5 | NP_611372.3 | 10 - 11 | 2850 | 76 | ||
| GenBank mRNA | AY071153.1 | - | 5 - 6 | 1597 | 76 | |
More Information
Set Membership: |
|

Mctp
Genomic Map