Assay Details
| Gene Symbol: Srpk79D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104190.4 | NP_001097660.2 | 1 - 2 | 1460 | 65 | |
| NM_001202232.3 | NP_001189161.1 | 1 - 2 | 1460 | 65 | ||
| NM_001259951.2 | NP_001246880.1 | 1 - 2 | 1460 | 65 | ||
| NM_001259952.2 | NP_001246881.1 | 1 - 2 | 1460 | 65 | ||
More Information
Set Membership: |
|

Srpk79D
Genomic Map