Assay Details
| Gene Symbol: Rpp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316642.1 | NP_001303571.1 | 1 - 2 | 250 | 60 | |
| NM_176680.4 | NP_788853.1 | 1 - 2 | 250 | 60 | ||
More Information
Set Membership: |
|

Rpp21
Genomic Map