Assay Details
| Gene Symbol: CG34417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272355.1 | NP_001259284.1 | 10 - 11 | 10100 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG34417| Gene Symbol: CG34417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272355.1 | NP_001259284.1 | 10 - 11 | 10100 | 81 | |
Set Membership: |
|