Assay Details
| Gene Symbol: CG33181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103438.2 | NP_001096908.1 | 3 - 4 | 1347 | 79 | |
| NM_001103441.2 | NP_001096911.1 | 3 - 4 | 1436 | 79 | ||
| NM_001103442.2 | NP_001096912.1 | 3 - 4 | 1555 | 79 | ||
| NM_001272405.1 | NP_001259334.1 | 4 - 5 | 1430 | 79 | ||
| NM_176712.3 | NP_788885.1 | 1 - 2 | 1031 | 79 | ||
| GenBank mRNA | BT120117.1 | - | 2 - 3 | 1416 | 79 | |
More Information
Set Membership: |
|

CG33181
Genomic Map