Assay Details
| Gene Symbol: CG33181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103438.2 | NP_001096908.1 | 5 - 6 | 2064 | 73 | |
| NM_001103440.2 | NP_001096910.1 | 6 - 7 | 1378 | 73 | ||
| NM_001103441.2 | NP_001096911.1 | 5 - 6 | 2153 | 73 | ||
| NM_001103442.2 | NP_001096912.1 | 5 - 6 | 2272 | 73 | ||
| NM_001272405.1 | NP_001259334.1 | 6 - 7 | 2147 | 73 | ||
| NM_001272406.1 | NP_001259335.1 | 6 - 7 | 1960 | 73 | ||
| NM_001298093.1 | NP_001285022.1 | 4 - 5 | 895 | 73 | ||
| NM_176712.3 | NP_788885.1 | 3 - 4 | 1748 | 73 | ||
| GenBank mRNA | BT120117.1 | - | 4 - 5 | 2133 | 73 | |
More Information
Set Membership: |
|

CG33181
Genomic Map