Assay Details
| Gene Symbol: CG33181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103438.2 | NP_001096908.1 | 7 - 8 | 2362 | 85 | |
| NM_001103440.2 | NP_001096910.1 | 8 - 9 | 1676 | 85 | ||
| NM_001103441.2 | NP_001096911.1 | 7 - 8 | 2451 | 85 | ||
| NM_001103442.2 | NP_001096912.1 | 7 - 8 | 2570 | 85 | ||
| NM_001272405.1 | NP_001259334.1 | 8 - 9 | 2445 | 85 | ||
| NM_001272406.1 | NP_001259335.1 | 8 - 9 | 2258 | 85 | ||
| NM_001298093.1 | NP_001285022.1 | 6 - 7 | 1193 | 85 | ||
| NM_176712.3 | NP_788885.1 | 5 - 6 | 2046 | 85 | ||
| GenBank mRNA | BT120117.1 | - | 6 - 7 | 2431 | 85 | |
More Information
Set Membership: |
|

CG33181
Genomic Map