Assay Details
| Gene Symbol: CG33181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103438.2 | NP_001096908.1 | 8 - 9 | 2570 | 56 | |
| NM_001103440.2 | NP_001096910.1 | 9 - 10 | 1884 | 56 | ||
| NM_001103441.2 | NP_001096911.1 | 8 - 9 | 2659 | 56 | ||
| NM_001103442.2 | NP_001096912.1 | 8 - 9 | 2778 | 56 | ||
| NM_001272405.1 | NP_001259334.1 | 9 - 10 | 2653 | 56 | ||
| NM_001272406.1 | NP_001259335.1 | 9 - 10 | 2466 | 56 | ||
| NM_001298093.1 | NP_001285022.1 | 7 - 8 | 1401 | 56 | ||
| NM_176712.3 | NP_788885.1 | 6 - 7 | 2254 | 56 | ||
| GenBank mRNA | BT120117.1 | - | 7 - 8 | 2639 | 56 | |
More Information
Set Membership: |
|

CG33181
Genomic Map