Assay Details
| Gene Symbol: inaE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144723.2 | NP_001138195.1 | 5 - 6 | 1116 | 68 | |
| NM_001144724.2 | NP_001138196.1 | 5 - 6 | 1116 | 68 | ||
| NM_001298301.1 | NP_001285230.1 | 5 - 6 | 1116 | 68 | ||
| NM_001298302.1 | NP_001285231.1 | 5 - 6 | 1116 | 68 | ||
| NM_176727.3 | NP_788900.1 | 5 - 6 | 1029 | 68 | ||
| NM_176728.3 | NP_788901.1 | 5 - 6 | 1116 | 68 | ||
| GenBank mRNA | AY122194.1 | - | 5 - 6 | 994 | 68 | |
| EU817579.1 | - | 5 - 6 | 994 | 68 | ||
| EU817580.1 | - | 5 - 6 | 994 | 68 | ||
More Information
Set Membership: |
|

inaE
Genomic Map