Assay Details
| Gene Symbol: hth | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057228.5 | NP_476576.1 | 13 - 14 | 2577 | 106 | |
| NM_057230.6 | NP_476578.3 | 13 - 14 | 2532 | 106 | ||
| GenBank mRNA | AF032865.1 | - | 13 - 14 | 1671 | 106 | |
| AF035825.1 | - | 13 - 14 | 2553 | 106 | ||
| AF036584.1 | - | 12 - 13 | 1713 | 106 | ||
More Information
Set Membership: |
|

hth
Genomic Map