Assay Details
| Gene Symbol: hth | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014613.3 | NP_001014613.2 | 5 - 6 | 2237 | 76 | |
| NM_001031999.2 | NP_001027170.1 | 4 - 5 | 1768 | 76 | ||
| NM_057228.5 | NP_476576.1 | 4 - 5 | 1768 | 76 | ||
| NM_057229.5 | NP_476577.3 | 4 - 5 | 1768 | 76 | ||
| NM_057230.6 | NP_476578.3 | 4 - 5 | 1768 | 76 | ||
| GenBank mRNA | AF032865.1 | - | 4 - 5 | 862 | 76 | |
| AF035825.1 | - | 4 - 5 | 1744 | 76 | ||
| AF036584.1 | - | 4 - 5 | 949 | 76 | ||
| BT024210.1 | - | 4 - 5 | 1587 | 76 | ||
| BT149862.1 | - | 5 - 6 | 1333 | 76 | ||
More Information
Set Membership: |
|

hth
Genomic Map