Assay Details
| Gene Symbol: AnxB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057255.3 | NP_476603.1 | 4 - 5 | 992 | 75 | |
| NM_206530.2 | NP_996252.1 | 4 - 5 | 1108 | 75 | ||
| GenBank mRNA | AF261718.1 | - | 4 - 5 | 1004 | 75 | |
| AY118504.1 | - | 4 - 5 | 1004 | 75 | ||
More Information
Set Membership: |
|

AnxB9
Genomic Map