Assay Details
| Gene Symbol: pyd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144556.2 | NP_001138028.1 | 8 - 9 | 4016 | 69 | |
| NM_001260069.2 | NP_001246998.1 | 9 - 10 | 4443 | 69 | ||
| NM_057349.7 | NP_476697.3 | 9 - 10 | 3769 | 69 | ||
| NM_169245.3 | NP_731291.2 | 8 - 9 | 3535 | 69 | ||
| NM_169246.3 | NP_731292.2 | 8 - 9 | 3535 | 69 | ||
| NM_170646.6 | NP_731293.3 | 10 - 11 | 5176 | 69 | ||
More Information
Set Membership: |
|

pyd
Genomic Map