Assay Details
| Gene Symbol: spn-B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057392.5 | NP_476740.1 | 3 - 4 | 717 | 99 | |
| GenBank mRNA | BT044133.1 | - | 3 - 4 | 704 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

spn-B| Gene Symbol: spn-B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057392.5 | NP_476740.1 | 3 - 4 | 717 | 99 | |
| GenBank mRNA | BT044133.1 | - | 3 - 4 | 704 | 99 | |
Set Membership: |
|