Assay Details
| Gene Symbol: spn-E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057393.4 | NP_476741.1 | 6 - 7 | 2616 | 58 | |
| GenBank mRNA | BT100306.1 | - | 6 - 7 | 2552 | 58 | |
More Information
Set Membership: |
|

spn-E
Genomic Map