Assay Details
| Gene Symbol: aurA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057401.4 | NP_476749.1 | 1 - 2 | 279 | 79 | |
| GenBank mRNA | AY061229.1 | - | 1 - 2 | 201 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aurA| Gene Symbol: aurA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057401.4 | NP_476749.1 | 1 - 2 | 279 | 79 | |
| GenBank mRNA | AY061229.1 | - | 1 - 2 | 201 | 79 | |
Set Membership: |
|