Assay Details
| Gene Symbol: bam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057452.4 | NP_476800.1 | 1 - 2 | 619 | 95 | |
| GenBank mRNA | BT044232.1 | - | 1 - 2 | 567 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

bam| Gene Symbol: bam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057452.4 | NP_476800.1 | 1 - 2 | 619 | 95 | |
| GenBank mRNA | BT044232.1 | - | 1 - 2 | 567 | 95 | |
Set Membership: |
|