Assay Details
| Gene Symbol: pll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276079.1 | NP_001263008.1 | 1 - 2 | 546 | 95 | |
| NM_057623.4 | NP_476971.1 | 1 - 2 | 546 | 95 | ||
| GenBank mRNA | BT056264.1 | - | 1 - 2 | 503 | 95 | |
| L08476.1 | - | 1 - 2 | 493 | 95 | ||
More Information
Set Membership: |
|

pll
Genomic Map