Assay Details
| Gene Symbol: Hmu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057811.4 | NP_477159.1 | 1 - 2 | 282 | 64 | |
| GenBank mRNA | BT133185.1 | - | 1 - 2 | 280 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmu| Gene Symbol: Hmu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057811.4 | NP_477159.1 | 1 - 2 | 282 | 64 | |
| GenBank mRNA | BT133185.1 | - | 1 - 2 | 280 | 64 | |
Set Membership: |
|