Assay Details
| Gene Symbol: Dad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014629.2 | NP_001014629.1 | 4 - 5 | 1815 | 69 | |
| NM_001382177.1 | NP_001369021.1 | 4 - 5 | 1815 | 69 | ||
| NM_057912.4 | NP_477260.1 | 4 - 5 | 1831 | 69 | ||
| NM_169740.2 | NP_732196.1 | 3 - 4 | 1614 | 69 | ||
| GenBank mRNA | BT016082.1 | - | 3 - 4 | 1614 | 69 | |
More Information
Set Membership: |
|

Dad
Genomic Map