Assay Details
| Gene Symbol: Dad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014629.2 | NP_001014629.1 | 6 - 7 | 2160 | 56 | |
| NM_001382177.1 | NP_001369021.1 | 6 - 7 | 2160 | 56 | ||
| NM_057912.4 | NP_477260.1 | 6 - 7 | 2176 | 56 | ||
| NM_169740.2 | NP_732196.1 | 5 - 6 | 1959 | 56 | ||
| GenBank mRNA | BT016082.1 | - | 5 - 6 | 1959 | 56 | |
More Information
Set Membership: |
|

Dad
Genomic Map