Assay Details
| Gene Symbol: Dad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014629.2 | NP_001014629.1 | 7 - 8 | 2319 | 65 | |
| NM_001382177.1 | NP_001369021.1 | 7 - 8 | 2319 | 65 | ||
| NM_057912.4 | NP_477260.1 | 7 - 8 | 2335 | 65 | ||
| NM_169740.2 | NP_732196.1 | 6 - 7 | 2118 | 65 | ||
| GenBank mRNA | BT016082.1 | - | 6 - 7 | 2118 | 65 | |
More Information
Set Membership: |
|

Dad
Genomic Map