Assay Details
| Gene Symbol: Abi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104315.2 | NP_001097785.1 | 1 - 2 | 504 | 67 | |
| NM_001260161.1 | NP_001247090.1 | 2 - 3 | 425 | 67 | ||
| NM_001260162.1 | NP_001247091.1 | 2 - 3 | 510 | 67 | ||
| NM_001275630.1 | NP_001262559.1 | 2 - 3 | 510 | 67 | ||
| NM_001275631.1 | NP_001262560.1 | 2 - 3 | 425 | 67 | ||
| NM_057915.5 | NP_477263.1 | 2 - 3 | 510 | 67 | ||
| GenBank mRNA | AF151115.1 | - | 2 - 3 | 488 | 67 | |
| AY051877.1 | - | 2 - 3 | 482 | 67 | ||
More Information
Set Membership: |
|

Abi
Genomic Map