Assay Details
| Gene Symbol: Dl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260264.2 | NP_001247193.1 | 2 - 3 | 1049 | 59 | |
| NM_057916.4 | NP_477264.1 | 2 - 3 | 1049 | 59 | ||
| NM_169853.3 | NP_732412.1 | 2 - 3 | 1049 | 59 | ||
| GenBank mRNA | BT003267.1 | - | 2 - 3 | 1021 | 59 | |
More Information
Set Membership: |
|

Dl
Genomic Map