Assay Details
| Gene Symbol: Dl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260264.2 | NP_001247193.1 | 5 - 6 | 1420 | 99 | |
| NM_057916.4 | NP_477264.1 | 5 - 6 | 1420 | 99 | ||
| NM_169853.3 | NP_732412.1 | 5 - 6 | 1420 | 99 | ||
| GenBank mRNA | BT003267.1 | - | 5 - 6 | 1392 | 99 | |
More Information
Set Membership: |
|

Dl
Genomic Map