Assay Details
| Gene Symbol: CRMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057959.3 | NP_477307.1 | 10 - 11 | 1353 | 99 | |
| NM_169070.2 | NP_730952.1 | 11 - 12 | 1592 | 99 | ||
| NM_169072.3 | NP_730954.2 | 10 - 11 | 1368 | 99 | ||
| GenBank mRNA | AF077837.1 | - | 10 - 11 | 1339 | 99 | |
| AF160900.1 | - | 10 - 11 | 1332 | 99 | ||
| AF465756.1 | - | 9 - 10 | 1235 | 99 | ||
| GU452673.1 | - | 10 - 11 | 1308 | 99 | ||
| GU452674.1 | - | 10 - 11 | 1320 | 99 | ||
| GU452675.1 | - | 10 - 11 | 1328 | 99 | ||
More Information
Set Membership: |
|

CRMP
Genomic Map