Assay Details
| Gene Symbol: CRMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057959.3 | NP_477307.1 | 2 - 3 | 214 | 62 | |
| NM_169070.2 | NP_730952.1 | 2 - 3 | 214 | 62 | ||
| NM_169072.3 | NP_730954.2 | 2 - 3 | 214 | 62 | ||
| GenBank mRNA | AF077837.1 | - | 2 - 3 | 200 | 62 | |
| AF160900.1 | - | 2 - 3 | 193 | 62 | ||
| AF465756.1 | - | 1 - 2 | 81 | 62 | ||
| GU452673.1 | - | 2 - 3 | 169 | 62 | ||
| GU452674.1 | - | 2 - 3 | 166 | 62 | ||
| GU452675.1 | - | 2 - 3 | 174 | 62 | ||
More Information
Set Membership: |
|

CRMP
Genomic Map