Assay Details
| Gene Symbol: CRMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057959.3 | NP_477307.1 | 7 - 8 | 747 | 73 | |
| NM_169070.2 | NP_730952.1 | 7 - 8 | 747 | 73 | ||
| NM_169072.3 | NP_730954.2 | 7 - 8 | 747 | 73 | ||
| GenBank mRNA | AF077837.1 | - | 7 - 8 | 733 | 73 | |
| AF160900.1 | - | 7 - 8 | 726 | 73 | ||
| AF465756.1 | - | 6 - 7 | 614 | 73 | ||
| GU452673.1 | - | 7 - 8 | 702 | 73 | ||
| GU452674.1 | - | 7 - 8 | 699 | 73 | ||
| GU452675.1 | - | 7 - 8 | 707 | 73 | ||
More Information
Set Membership: |
|

CRMP
Genomic Map