Assay Details
| Gene Symbol: CRMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057959.3 | NP_477307.1 | 9 - 10 | 1110 | 86 | |
| NM_169070.2 | NP_730952.1 | 10 - 11 | 1349 | 86 | ||
| GenBank mRNA | AF077837.1 | - | 9 - 10 | 1096 | 86 | |
| AF160900.1 | - | 9 - 10 | 1089 | 86 | ||
| GU452673.1 | - | 9 - 10 | 1065 | 86 | ||
More Information
Set Membership: |
|

CRMP
Genomic Map