Assay Details
| Gene Symbol: Mkk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170080.2 | NP_001163551.1 | 3 - 4 | 1298 | 92 | |
| NM_001275446.1 | NP_001262375.1 | 3 - 4 | 1298 | 92 | ||
| NM_058005.5 | NP_477353.1 | 3 - 4 | 1298 | 92 | ||
| GenBank mRNA | AY070600.1 | - | 3 - 4 | 1296 | 92 | |
More Information
Set Membership: |
|

Mkk4
Genomic Map