Assay Details
| Gene Symbol: Pcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058012.4 | NP_477360.2 | 2 - 3 | 343 | 61 | |
| GenBank mRNA | BT088995.1 | - | 2 - 3 | 343 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcd| Gene Symbol: Pcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058012.4 | NP_477360.2 | 2 - 3 | 343 | 61 | |
| GenBank mRNA | BT088995.1 | - | 2 - 3 | 343 | 61 | |
Set Membership: |
|